Assay Details
Gene Symbol: DLA-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014379.1 | NP_001014401.1 | 2 | 261 | 143 | |
GenBank mRNA | LC130528.1 | - | 2 | 261 | 143 | |
LC130529.1 | - | 2 | 261 | 143 | ||
LC130530.1 | - | 2 | 261 | 143 | ||
LC130531.1 | - | 2 | 261 | 143 | ||
LC130532.1 | - | 2 | 261 | 143 | ||
LC130533.1 | - | 2 | 261 | 143 | ||
LC130534.1 | - | 2 | 261 | 143 | ||
LC130535.1 | - | 2 | 261 | 143 | ||
LC130536.1 | - | 2 | 261 | 143 | ||
LC130537.1 | - | 2 | 261 | 143 | ||
LC130538.1 | - | 2 | 261 | 143 | ||
LC130539.1 | - | 2 | 261 | 143 | ||
LC130541.1 | - | 2 | 261 | 143 | ||
LC130543.1 | - | 2 | 261 | 143 | ||
LC130544.1 | - | 2 | 261 | 143 | ||
LC130545.1 | - | 2 | 261 | 143 | ||
LC130547.1 | - | 2 | 261 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
