Assay Details
| Gene Symbol: GJC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020812.2 | NP_001018648.1 | 1124 | 88 | ||
| XM_005624343.3 | XP_005624400.1 | 1396 | 88 | |||
| XM_005624344.3 | XP_005624401.1 | 1538 | 88 | |||
More Information
Set Membership: |
|

GJC1
Genomic Map