Assay Details
| Gene Symbol: AANAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077437.1 | NP_001070905.1 | 159 | 57 | ||
| XM_005623964.3 | XP_005624021.1 | 424 | 57 | |||
| GenBank mRNA | DQ336162.1 | - | 159 | 57 | ||
More Information
Set Membership: |
|

AANAT
Genomic Map