Assay Details
Gene Symbol: SMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097558.1 | NP_001091027.1 | 9 - 10 | 969 | 75 | |
XM_005627008.2 | XP_005627065.1 | 8 - 9 | 1033 | 75 | ||
XM_005627009.2 | XP_005627066.1 | 9 - 10 | 973 | 75 | ||
XM_014118014.2 | XP_013973489.1 | 10 - 11 | 1163 | 75 | ||
XM_014118015.2 | XP_013973490.1 | 10 - 11 | 1021 | 75 | ||
XM_022425702.1 | XP_022281410.1 | 10 - 11 | 1078 | 75 | ||
XM_022425703.1 | XP_022281411.1 | 7 - 8 | 951 | 75 | ||
XM_022425704.1 | XP_022281412.1 | 10 - 11 | 1018 | 75 | ||
GenBank mRNA | DQ999010.1 | - | 9 - 10 | 969 | 75 | |
EF116598.1 | - | 8 - 9 | 888 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
