Assay Details
| Gene Symbol: B3GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048137.1 | NP_001041602.1 | 867 | 153 | ||
| GenBank mRNA | AM235753.1 | - | 867 | 153 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3GALT6| Gene Symbol: B3GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048137.1 | NP_001041602.1 | 867 | 153 | ||
| GenBank mRNA | AM235753.1 | - | 867 | 153 | ||
Set Membership: |
|