Assay Details
| Gene Symbol: CPT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625702.3 | XP_005625759.1 | 2 - 3 | 378 | 63 | |
| XM_022424242.1 | XP_022279950.1 | 2 - 3 | 384 | 63 | ||
| XM_022424243.1 | XP_022279951.1 | 2 - 3 | 384 | 63 | ||
| XM_022424244.1 | XP_022279952.1 | 2 - 3 | 379 | 63 | ||
| XM_538305.6 | XP_538305.2 | 2 - 3 | 374 | 63 | ||
More Information
Set Membership: |
|

CPT1B
Genomic Map