Assay Details
| Gene Symbol: TNFSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130836.1 | NP_001124308.1 | 4 - 5 | 407 | 87 | |
| GenBank mRNA | EU036633.1 | - | 4 - 5 | 407 | 87 | |
More Information
Set Membership: |
|

TNFSF10
Genomic Map