Assay Details
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130838.1 | NP_001124310.1 | 2 - 3 | 290 | 60 | |
XM_005628844.3 | XP_005628901.1 | 3 - 4 | 690 | 60 | ||
XM_022427669.1 | XP_022283377.1 | 3 - 4 | 632 | 60 | ||
GenBank mRNA | EU038071.1 | - | 2 - 3 | 290 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
