Assay Details
Gene Symbol: TAS2R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145499.1 | NP_001138971.1 | 846 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
