Assay Details
| Gene Symbol: SIRPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634881.3 | XP_005634938.1 | 5 - 6 | 1240 | 71 | |
| XM_014106991.2 | XP_013962466.1 | 4 - 5 | 1147 | 71 | ||
| XM_014106992.2 | XP_013962467.1 | 4 - 5 | 1147 | 71 | ||
More Information
Set Membership: |
|

SIRPA
Genomic Map