Assay Details
| Gene Symbol: OBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190926.1 | NP_001177855.1 | 1 - 2 | 73 | 91 | |
| XM_005640968.2 | XP_005641025.1 | 1 - 2 | 133 | 91 | ||
| GenBank mRNA | GU132996.1 | - | 1 - 2 | 73 | 91 | |
| KF192077.2 | - | 1 - 2 | 73 | 91 | ||
More Information
Set Membership: |
|

OBP
Genomic Map