Assay Details
| Gene Symbol: TNFSF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205169.1 | NP_001192098.1 | 3 - 4 | 408 | 65 | |
| GenBank mRNA | DN371994.1 | - | 3 - 4 | 408 | 65 | |
| EU909456.1 | - | 3 - 4 | 383 | 65 | ||
More Information
Set Membership: |
|

TNFSF13
Genomic Map