Assay Details
| Gene Symbol: UBE2V1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205096.1 | NP_001192025.1 | 4 | 1054 | 77 | |
| XM_014106824.2 | XP_013962299.1 | 4 | 1098 | 77 | ||
| XM_022408797.1 | XP_022264505.1 | 3 | 995 | 77 | ||
| GenBank mRNA | DN867801.1 | - | 1 | 503 | 77 | |
More Information
Set Membership: |
|

UBE2V1
Genomic Map