Assay Details
| Gene Symbol: NDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252172.1 | NP_001239101.1 | 5 - 6 | 765 | 71 | |
| XM_005621297.1 | XP_005621354.1 | 6 - 7 | 783 | 71 | ||
| XM_022419473.1 | XP_022275181.1 | 6 - 7 | 946 | 71 | ||
| GenBank mRNA | DN381130.1 | - | 1 - 2 | 180 | 71 | |
More Information
Set Membership: |
|

NDE1
Genomic Map