Assay Details
Gene Symbol: MAPRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251946.1 | NP_001238875.1 | 4 - 5 | 547 | 79 | |
GenBank mRNA | DN267006.1 | - | 4 - 5 | 547 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
