Assay Details
| Gene Symbol: TAF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252038.1 | NP_001238967.1 | 3 - 4 | 261 | 87 | |
| GenBank mRNA | DN372994.1 | - | 3 - 4 | 261 | 87 | |
More Information
Set Membership: |
|

TAF13
Genomic Map