Assay Details
| Gene Symbol: RPS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252045.1 | NP_001238974.1 | 3 - 4 | 384 | 74 | |
| XM_005627054.1 | XP_005627111.1 | 3 - 4 | 552 | 74 | ||
| GenBank mRNA | CO633541.1 | - | 1 | 224 | 74 | |
More Information
Set Membership: |
|

RPS10
Genomic Map