Assay Details
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252323.1 | NP_001239252.1 | 2 - 3 | 467 | 86 | |
XM_005617237.3 | XP_005617294.1 | 2 - 3 | 540 | 86 | ||
GenBank mRNA | DN379330.1 | - | 2 - 3 | 465 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
