Assay Details
| Gene Symbol: CKLF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252340.1 | NP_001239269.1 | 1 - 2 | 78 | 103 | |
| XM_005620618.3 | XP_005620675.1 | 1 - 2 | 166 | 103 | ||
More Information
Set Membership: |
|

CKLF
Genomic Map