Assay Details
| Gene Symbol: CKLF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252340.1 | NP_001239269.1 | 2 - 3 | 232 | 70 | |
| XM_005620618.3 | XP_005620675.1 | 2 - 3 | 320 | 70 | ||
More Information
Set Membership: |
|

CKLF
Genomic Map