Assay Details
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252340.1 | NP_001239269.1 | 3 - 4 | 329 | 84 | |
XM_005620620.3 | XP_005620677.1 | 2 - 3 | 253 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
