Assay Details
Gene Symbol: CMTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252345.1 | NP_001239274.1 | 1 - 2 | 486 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
