Assay Details
| Gene Symbol: RPL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252411.1 | NP_001239340.1 | 2 - 3 | 154 | 76 | |
| GenBank mRNA | GR886597.1 | - | 2 - 3 | 154 | 76 | |
More Information
Set Membership: |
|

RPL14
Genomic Map