Assay Details
| Gene Symbol: DYNLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252419.1 | NP_001239348.1 | 3 - 4 | 318 | 99 | |
| XM_005634683.3 | XP_005634740.1 | 2 - 3 | 266 | 99 | ||
| GenBank mRNA | GR886117.1 | - | 3 - 4 | 318 | 99 | |
More Information
Set Membership: |
|

DYNLRB1
Genomic Map