Assay Details
| Gene Symbol: KRT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253741.1 | NP_001240670.1 | 1139 | 70 | ||
| GenBank mRNA | DN872732.1 | - | 372 | 70 | ||
| DN873542.1 | - | 544 | 70 | |||
| DN873543.1 | - | 587 | 70 | |||
More Information
Set Membership: |
|

KRT14
Genomic Map