Assay Details
Gene Symbol: RAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194984.1 | NP_001181913.1 | 1 | 629 | 88 | |
GenBank mRNA | BU745248.1 | - | 1 | 668 | 88 | |
Z11922.1 | - | 629 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
