Assay Details
| Gene Symbol: LOC478701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_535868.6 | XP_535868.3 | 2 | 216 | 110 | |
| Gene Symbol: LOC102152648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110500.2 | XP_013965975.2 | 3 | 190 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_535868.6 transcripts.
Set Membership: |
|

LOC478701
Genomic Map