Assay Details
| Gene Symbol: CXCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640640.2 | XP_005640697.1 | 1 - 2 | 76 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005640640.2 transcripts.
Set Membership: |
|

CXCR1
Genomic Map