Assay Details
| Gene Symbol: CXCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640416.2 | XP_005640473.1 | 2 - 3 | 245 | 97 | |
| XM_005640417.2 | XP_005640474.1 | 2 - 3 | 327 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005640416.2, XM_005640417.2 transcripts.
Set Membership: |
|

CXCR2
Genomic Map