Assay Details
| Gene Symbol: MIR758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049513.1 | 1 | 44 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | VRK serine/threonine kinase 1 | VRK1 | -1.5mb | Sense | Chr. 8 - 65444404 - 65551348 on NCBI Build CanFam3.1 | |
| Downstream | BAF chromatin remodeling complex subunit BCL11B | BCL11B | 524.8kb | Anti-sense | Chr. 8 - 67622592 - 67713922 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR758
Genomic Map