Assay Details
| Gene Symbol: MIR449A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049215.1 | 1 | 45 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myosin IIIA | MYO3A | -179.2kb | Sense | Chr. 2 - 7455125 - 7695483 on NCBI Build CanFam3.1 | |
| Downstream | G protein-coupled receptor 158 | GPR158 | 95.0kb | Sense | Chr. 2 - 7969781 - 8379657 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR449A
Genomic Map