Assay Details
Gene Symbol: MIR301A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049441.1 | 1 | 45 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATP binding cassette subfamily A member 9 | ABCA9 | Sense | Chr. 9 - 15515462 - 15574890 on NCBI Build CanFam3.1 | ||
Downstream | ATP binding cassette subfamily A member 6 | ABCA6 | 52.1kb | Sense | Chr. 9 - 15587837 - 15648639 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
