Assay Details
Gene Symbol: MIR218-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049324.1 | 1 | 31 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | retinal G protein coupled receptor | RGR | -18.2kb | Sense | Chr. 4 - 32489964 - 32499393 on NCBI Build CanFam3.1 | |
Downstream | coiled-coil serine rich protein 2 | CCSER2 | FAM190B | 26.0kb | Sense | Chr. 4 - 32543705 - 32733149 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
