Assay Details
| Gene Symbol: MIR195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049332.1 | 1 | 34 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 5 C17orf49 homolog | C5H17orf49 | -3.0 | Anti-sense | Chr. 5 - 32042545 - 32045347 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR195
Genomic Map