Assay Details
Gene Symbol: MIR488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049492.1 | 1 | 43 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | translation initiation factor IF-2-like | LOC106558992 | -136.7kb | Sense | Chr. 7 - 20552295 - 20557194 on NCBI Build CanFam3.1 | |
Downstream |
ABL proto-oncogene 2, non-receptor tyrosine kinase |
ABL2 | 2.7kb | Sense | Chr. 7 - 20696659 - 20735362 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
