Assay Details
| Gene Symbol: MIR653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049512.1 | 1 | 41 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calcitonin receptor | CALCR | Ct-R | Sense | Chr. 14 - 18923116 - 19074904 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR653
Genomic Map