Assay Details
Gene Symbol: MIR8822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128763.1 | 1 | 57 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transient receptor potential cation channel subfamily C member 4 | TRPC4 | Sense | Chr. 25 - 2927347 - 3133935 on NCBI Build CanFam3.1 | ||
Downstream | periostin | POSTN | 78.5kb | Sense | Chr. 25 - 3167319 - 3208970 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
