Assay Details
| Gene Symbol: MIR8877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128833.1 | 1 | 23 | 53 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein kinase C beta | PRKCB | PRKCB1 | Anti-sense | Chr. 6 - 21742970 - 22064360 on NCBI Build CanFam3.1 | |
| Downstream | calcineurin like EF-hand protein 2 | CHP2 | 107.1kb | Anti-sense | Chr. 6 - 22127422 - 22129637 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8877
Genomic Map