Assay Details
| Gene Symbol: MIR27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049270.1 | 1 | 25 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TAFA chemokine like family member 1 | TAFA1 | FAM19A1 | Anti-sense | Chr. 20 - 23128171 - 23621402 on NCBI Build CanFam3.1 | |
| Downstream | succinate-CoA ligase GDP-forming subunit beta | SUCLG2 | 481.1kb | Sense | Chr. 20 - 23915149 - 24167740 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR27A
Genomic Map