Assay Details
Gene Symbol: MIR708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049273.1 | 1 | 36 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 181 member B | FAM181B | -3.2mb | Sense | Chr. 21 - 16355669 - 16357654 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
