Assay Details
| Gene Symbol: MIR130B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049292.1 | 1 | 32 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 132B | TMEM132B | -124.1kb | Anti-sense | Chr. 26 - 4446667 - 4683425 on NCBI Build CanFam3.1 | |
| Downstream | protein cornichon homolog 4 | LOC102155917 | 33.3kb | Anti-sense | Chr. 26 - 4840822 - 4841166 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR130B
Genomic Map