Assay Details
Gene Symbol: MIR219-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049376.1 | 1 | 36 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | forkhead box protein L2-like | LOC111097301 | Anti-sense | Chr. 9 - 7319682 - 7380459 on NCBI Build CanFam3.1 | ||
Downstream | uncharacterized LOC100856617 | LOC100856617 | 54.3kb | Anti-sense | Chr. 9 - 7418390 - 7498630 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
