Assay Details
Gene Symbol: MIR8803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128743.1 | 1 | 86 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PDZ domain containing ring finger 3 | PDZRN3 | -286.1kb | Anti-sense | Chr. 20 - 18621960 - 18859620 on NCBI Build CanFam3.1 | |
Downstream | glucoside xylosyltransferase 2 | GXYLT2 | 39.0kb | Sense | Chr. 20 - 19184892 - 19253144 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
