Assay Details
| Gene Symbol: MIR8806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128746.1 | 1 | 90 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC111091887 | LOC111091887 | -103.6kb | Anti-sense | Chr. 23 - 12126745 - 12131244 on NCBI Build CanFam3.1 | |
| Downstream | dynein cytoplasmic 1 light intermediate chain 1 | DYNC1LI1 | 575.0 | Anti-sense | Chr. 23 - 12235539 - 12283851 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR8806
Genomic Map