Assay Details
Gene Symbol: MIR8883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128842.1 | 1 | 101 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sterile alpha motif domain containing 4A | SAMD4A | -869.0 | Sense | Chr. 8 - 30476425 - 30687240 on NCBI Build CanFam3.1 | |
Downstream | GTP cyclohydrolase 1 | GCH1 | 45.2kb | Anti-sense | Chr. 8 - 30733422 - 30784067 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
