Assay Details
| Gene Symbol: EIF3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251940.1 | NP_001238869.1 | 12 - 13 | 1680 | 65 | |
| GenBank mRNA | DN408493.1 | - | 1 | 457 | 65 | |
More Information
Set Membership: |
|

EIF3L
Genomic Map