Assay Details
| Gene Symbol: DEFB126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284438.1 | NP_001271367.1 | 1 - 2 | 57 | 94 | |
| GenBank mRNA | DQ012009.1 | - | 1 - 2 | 57 | 94 | |
More Information
Set Membership: |
|

DEFB126
Genomic Map