Assay Details
| Gene Symbol: Sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495418.2 | XP_003495466.1 | 3 - 4 | 423 | 75 | |
| XM_007640868.1 | XP_007639058.1 | 3 - 4 | 527 | 75 | ||
| XM_007640874.1 | XP_007639064.1 | 3 - 4 | 434 | 75 | ||
| GenBank mRNA | JI876865.1 | - | 3 - 4 | 418 | 75 | |
| JP040342.1 | - | 3 - 4 | 421 | 75 | ||
More Information
Set Membership: |
|

Sec22a
Genomic Map