Assay Details
Gene Symbol: Ptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641702.1 | XP_007639892.1 | 2 - 3 | 288 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
