Assay Details
Gene Symbol: Ptchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640124.1 | XP_007638314.1 | 2 - 3 | 714 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
